Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06598191_mH
          See other ARMH1 GE Assays ›
          Gene Symbol:
          ARMH1
          Gene Name:
          armadillo like helical domain containing 1
          Gene Aliases:
          C1orf228, NCRNA00082, p40
          Chromosome Location:
          Chr.1: 44674714 - 44725591 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06598191_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ARMH1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145636.2 NP_001139108.1 2 - 3 381 79
          XM_006710603.3 XP_006710666.1 3 - 4 645 79
          XM_006710604.3 XP_006710667.1 3 - 4 633 79
          XM_011541340.2 XP_011539642.1 4 - 5 769 79
          XM_011541341.2 XP_011539643.1 2 - 3 381 79
          XM_011541343.2 XP_011539645.1 3 - 4 505 79
          XM_011541345.2 XP_011539647.1 3 - 4 579 79
          XM_011541349.3 XP_011539651.1 2 - 3 315 79
          XM_017001142.2 XP_016856631.1 2 - 3 381 79
          XM_017001143.2 XP_016856632.1 2 - 3 381 79
          XM_017001144.2 XP_016856633.1 2 - 3 381 79
          XM_017001145.2 XP_016856634.1 2 - 3 381 79
          XM_017001146.3 XP_016856635.1 2 - 3 381 79
          XM_017001147.2 XP_016856636.1 2 - 3 381 79
          XM_047419337.1 XP_047275293.1 3 - 4 515 79
          XM_047419352.1 XP_047275308.1 3 - 4 497 79
          XM_047419353.1 XP_047275309.1 2 - 3 381 79
          XM_047419358.1 XP_047275314.1 2 - 3 381 79
          XM_047419362.1 XP_047275318.1 2 - 3 381 79
          XM_047419367.1 XP_047275323.1 2 - 3 381 79
          XM_047419369.1 XP_047275325.1 2 - 3 381 79
          XM_047419370.1 XP_047275326.1 2 - 3 381 79
          XM_047419371.1 XP_047275327.1 2 - 3 381 79
          GenBank mRNA BC026115.1 - 2 - 3 377 79

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_011541349.3 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline