Search
Search

SPIN2B| Gene Symbol: SPIN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006681.2 | NP_001006682.1 | 1 - 2 | 308 | 100 | |
| NM_001282461.2 | NP_001269390.1 | 1 - 2 | 308 | 100 | ||
| GenBank mRNA | BC071974.1 | - | 1 - 2 | 238 | 100 | |
Set Membership: |
|