Search
Search

BTNL10| Gene Symbol: BTNL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287262.1 | NP_001274191.1 | 2 - 3 | 460 | 83 | |
| XM_011544015.2 | XP_011542317.1 | 2 - 3 | 1668 | 83 | ||
| XM_016999986.1 | XP_016855475.1 | 1 - 2 | 2390 | 83 | ||
| XM_016999987.1 | XP_016855476.1 | 2 - 3 | 1668 | 83 | ||
| GenBank mRNA | DY654633.1 | - | 2 - 3 | 421 | 83 | |
Set Membership: |
|