Search
Search

GANAB| Gene Symbol: GANAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285075.1 | NP_001272004.1 | 23 - 24 | 2729 | 55 | |
| XM_005577578.2 | XP_005577635.1 | 24 - 25 | 2911 | 55 | ||
| GenBank mRNA | AB072762.1 | - | 12 - 13 | 1182 | 55 | |
| AB169857.1 | - | 23 - 24 | 2730 | 55 | ||
| AB173690.1 | - | 11 - 12 | 1204 | 55 | ||
Set Membership: |
|