Search
Search

CEP78| Gene Symbol: CEP78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581980.2 | XP_005582037.1 | 4 - 5 | 879 | 69 | |
| XM_005581981.2 | XP_005582038.1 | 4 - 5 | 879 | 69 | ||
| XM_015437304.1 | XP_015292790.1 | 4 - 5 | 630 | 69 | ||
| XM_015437305.1 | XP_015292791.1 | 4 - 5 | 630 | 69 | ||
| XM_015437306.1 | XP_015292792.1 | 4 - 5 | 630 | 69 | ||
| XM_015437307.1 | XP_015292793.1 | 4 - 5 | 864 | 69 | ||
| XM_015437308.1 | XP_015292794.1 | 4 - 5 | 630 | 69 | ||
| XM_015437309.1 | XP_015292795.1 | 4 - 5 | 630 | 69 | ||
| XM_015437311.1 | XP_015292797.1 | 4 - 5 | 630 | 69 | ||
| XM_015437312.1 | XP_015292798.1 | 4 - 5 | 630 | 69 | ||
| XM_015437313.1 | XP_015292799.1 | 4 - 5 | 630 | 69 | ||
| XM_015437314.1 | XP_015292800.1 | 4 - 5 | 630 | 69 | ||
| XM_015437315.1 | XP_015292801.1 | 4 - 5 | 630 | 69 | ||
Set Membership: |
|