Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mf00405524_m1
          See other LIMCH1 GE Assays ›
          Gene Symbol:
          LIMCH1
          Gene Name:
          LIM and calponin homology domains 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 41105870 - 41450447 on Build M_fas_5.0
          Species:
          Macaca fascicularis
          Species Specific ID (Flybase ID):
          -
          Assay ID Mf00405524_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LIMCH1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005554755.2 XP_005554812.1 8 - 9 740 74
          XM_005554756.2 XP_005554813.1 8 - 9 740 74
          XM_005554757.2 XP_005554814.1 8 - 9 740 74
          XM_005554758.2 XP_005554815.1 7 - 8 719 74
          XM_005554759.2 XP_005554816.1 8 - 9 740 74
          XM_005554760.2 XP_005554817.1 8 - 9 740 74
          XM_005554761.2 XP_005554818.1 8 - 9 740 74
          XM_005554762.2 XP_005554819.1 8 - 9 740 74
          XM_005554763.2 XP_005554820.1 8 - 9 740 74
          XM_005554764.2 XP_005554821.1 8 - 9 740 74
          XM_005554765.2 XP_005554822.1 5 - 6 1064 74
          XM_005554766.2 XP_005554823.1 8 - 9 740 74
          XM_005554767.2 XP_005554824.1 8 - 9 626 74
          XM_005554769.2 XP_005554826.1 8 - 9 740 74
          XM_005554770.2 XP_005554827.1 8 - 9 740 74
          XM_005554771.2 XP_005554828.1 8 - 9 740 74
          XM_005554772.1 XP_005554829.1 7 - 8 719 74
          XM_005554773.2 XP_005554830.1 8 - 9 740 74
          XM_005554774.1 XP_005554831.1 7 - 8 719 74
          XM_005554775.1 XP_005554832.1 7 - 8 719 74
          XM_005554776.2 XP_005554833.1 2 - 3 919 74
          XM_005554777.2 XP_005554834.1 2 - 3 914 74
          XM_005554778.2 XP_005554835.1 2 - 3 574 74
          XM_015450296.1 XP_015305782.1 8 - 9 740 74
          XM_015450297.1 XP_015305783.1 8 - 9 740 74
          XM_015450298.1 XP_015305784.1 2 - 3 4120 74
          XM_015450299.1 XP_015305785.1 7 - 8 2959 74
          XM_015450300.1 XP_015305786.1 8 - 9 1514 74
          XM_015450301.1 XP_015305787.1 5 - 6 773 74
          XM_015450302.1 XP_015305788.1 8 - 9 740 74
          XM_015450303.1 XP_015305789.1 7 - 8 719 74
          XM_015450304.1 XP_015305790.1 8 - 9 740 74
          XM_015450305.1 XP_015305791.1 8 - 9 740 74
          XM_015450306.1 XP_015305792.1 2 - 3 933 74
          XM_015450307.1 XP_015305793.1 2 - 3 921 74
          XM_015450308.1 XP_015305794.1 2 - 3 920 74
          XM_015450309.1 XP_015305795.1 7 - 8 2942 74

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Probe spans exons

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline