Search
Search

CFAP57| Gene Symbol: CFAP57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543691.1 | XP_005543748.1 | 12 - 13 | 2236 | 68 | |
| XM_005543692.2 | XP_005543749.1 | 12 - 13 | 2236 | 68 | ||
| XM_015437254.1 | XP_015292740.1 | 11 - 12 | 2373 | 68 | ||
| XM_015437256.1 | XP_015292742.1 | 12 - 13 | 2236 | 68 | ||
Set Membership: |
|