Search
Search

CCDC58| Gene Symbol: CCDC58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015446357.1 | XP_015301843.1 | 2 - 3 | 238 | 69 | |
| XM_015446358.1 | XP_015301844.1 | 2 - 3 | 202 | 69 | ||
| XM_015446359.1 | XP_015301845.1 | 2 - 3 | 387 | 69 | ||
| XM_015446360.1 | XP_015301846.1 | 2 - 3 | 238 | 69 | ||
| XM_015446362.1 | XP_015301848.1 | 2 - 3 | 202 | 69 | ||
Set Membership: |
|