Search
Search

CHRND| Gene Symbol: CHRND | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574618.2 | XP_005574675.1 | 9 - 10 | 1623 | 78 | |
| XM_005574619.2 | XP_005574676.1 | 10 - 11 | 1539 | 78 | ||
| XM_005574620.2 | XP_005574677.1 | 10 - 11 | 1494 | 78 | ||
| XM_005574621.2 | XP_005574678.1 | 11 - 12 | 1431 | 78 | ||
| XM_015432969.1 | XP_015288455.1 | 6 - 7 | 793 | 78 | ||
Set Membership: |
|