Search
Search

OS9| Gene Symbol: OS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571371.2 | XP_005571428.1 | 1 - 2 | 373 | 78 | |
| XM_005571372.2 | XP_005571429.1 | 1 - 2 | 373 | 78 | ||
| XM_005571373.2 | XP_005571430.1 | 1 - 2 | 373 | 78 | ||
| XM_005571374.2 | XP_005571431.1 | 1 - 2 | 373 | 78 | ||
| XM_005571375.2 | XP_005571432.1 | 1 - 2 | 373 | 78 | ||
| XM_005571376.2 | XP_005571433.1 | 1 - 2 | 373 | 78 | ||
| XM_005571377.2 | XP_005571434.1 | 1 - 2 | 373 | 78 | ||
| XM_015431125.1 | XP_015286611.1 | 1 - 2 | 373 | 78 | ||
| XM_015431126.1 | XP_015286612.1 | 1 - 2 | 373 | 78 | ||
| GenBank mRNA | AB168638.1 | - | 196 | 78 | ||
Set Membership: |
|