Search
Search

MON2| Gene Symbol: MON2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571421.2 | XP_005571478.1 | 29 - 30 | 4618 | 102 | |
| XM_005571422.2 | XP_005571479.1 | 28 - 29 | 4615 | 102 | ||
| XM_005571427.2 | XP_005571484.1 | 28 - 29 | 4040 | 102 | ||
| XM_015431142.1 | XP_015286628.1 | 29 - 30 | 4621 | 102 | ||
| XM_015431143.1 | XP_015286629.1 | 30 - 31 | 4446 | 102 | ||
Set Membership: |
|