Search
Search

HIF1A| Gene Symbol: HIF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283896.1 | NP_001270825.1 | 13 - 14 | 2498 | 94 | |
| XM_005561440.2 | XP_005561497.1 | 13 - 14 | 2607 | 94 | ||
| XM_015453208.1 | XP_015308694.1 | 12 - 13 | 2484 | 94 | ||
| GenBank mRNA | AB169332.1 | - | 13 - 14 | 2498 | 94 | |
| AB174619.1 | - | 4 - 5 | 899 | 94 | ||
Set Membership: |
|