Search
Search

RNF17| Gene Symbol: RNF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585492.2 | XP_005585549.1 | 24 - 25 | 3341 | 73 | |
| XM_005585493.2 | XP_005585550.1 | 24 - 25 | 3341 | 73 | ||
| XM_005585495.2 | XP_005585552.1 | 24 - 25 | 3379 | 73 | ||
| XM_005585498.2 | XP_005585555.1 | 24 - 25 | 3255 | 73 | ||
| XM_015439073.1 | XP_015294559.1 | 24 - 25 | 3293 | 73 | ||
| XM_015439074.1 | XP_015294560.1 | 21 - 22 | 3043 | 73 | ||
| GenBank mRNA | AB071045.1 | - | 10 - 11 | 1435 | 73 | |
| AB179302.1 | - | 20 - 21 | 2862 | 73 | ||
Set Membership: |
|