Search
Search

LTBP4| Gene Symbol: LTBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589287.2 | XP_005589344.1 | 8 - 9 | 1088 | 97 | |
| XM_005589288.2 | XP_005589345.1 | 8 - 9 | 1088 | 97 | ||
| XM_005589290.2 | XP_005589347.1 | 8 - 9 | 1088 | 97 | ||
| XM_005589291.2 | XP_005589348.1 | 8 - 9 | 1088 | 97 | ||
| XM_015441540.1 | XP_015297026.1 | 8 - 9 | 1636 | 97 | ||
Set Membership: |
|