Search
Search

SYTL5| Gene Symbol: SYTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593281.2 | XP_005593338.1 | 16 - 17 | 2467 | 64 | |
| XM_005593282.2 | XP_005593339.1 | 16 - 17 | 3863 | 64 | ||
| XM_005593285.2 | XP_005593342.1 | 16 - 17 | 2382 | 64 | ||
| XM_015443561.1 | XP_015299047.1 | 16 - 17 | 2379 | 64 | ||
| XM_015443562.1 | XP_015299048.1 | 15 - 16 | 2316 | 64 | ||
| XM_015443563.1 | XP_015299049.1 | 15 - 16 | 2247 | 64 | ||
| XM_015443564.1 | XP_015299050.1 | 13 - 14 | 2022 | 64 | ||
Set Membership: |
|