Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mf00948093_g1
          See other HUWE1 GE Assays ›
          Gene Symbol:
          HUWE1
          Gene Name:
          HECT, UBA and WWE domain containing E3 ubiquitin protein ligase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 52605172 - 52757970 on Build M_fas_5.0
          Species:
          Macaca fascicularis
          Species Specific ID (Flybase ID):
          -
          Assay ID Mf00948093_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HUWE1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005593658.2 XP_005593715.1 30 - 31 3768 108
          XM_005593659.2 XP_005593716.1 30 - 31 8465 108
          XM_015443388.1 XP_015298874.1 28 - 29 3675 108
          XM_015443389.1 XP_015298875.1 28 - 29 3654 108
          XM_015443390.1 XP_015298876.1 30 - 31 4483 108
          XM_015443391.1 XP_015298877.1 31 - 32 4430 108
          XM_015443392.1 XP_015298878.1 31 - 32 4210 108
          XM_015443393.1 XP_015298879.1 31 - 32 4426 108
          XM_015443394.1 XP_015298880.1 32 - 33 4264 108
          XM_015443395.1 XP_015298881.1 31 - 32 4206 108
          XM_015443396.1 XP_015298882.1 31 - 32 4027 108
          XM_015443397.1 XP_015298883.1 30 - 31 4086 108
          XM_015443399.1 XP_015298885.1 30 - 31 4294 108
          XM_015443400.1 XP_015298886.1 31 - 32 4128 108
          XM_015443401.1 XP_015298887.1 30 - 31 3922 108
          XM_015443402.1 XP_015298888.1 30 - 31 5736 108
          XM_015443403.1 XP_015298889.1 31 - 32 8730 108
          XM_015443404.1 XP_015298890.1 30 - 31 3881 108
          XM_015443405.1 XP_015298891.1 29 - 30 3760 108
          XM_015443406.1 XP_015298892.1 29 - 30 3936 108
          XM_015443407.1 XP_015298893.1 28 - 29 3648 108
          XM_015443408.1 XP_015298894.1 28 - 29 3675 108
          XM_015443409.1 XP_015298895.1 28 - 29 3675 108
          XM_015443410.1 XP_015298896.1 28 - 29 3675 108
          XM_015443411.1 XP_015298897.1 28 - 29 3675 108
          XM_015443412.1 XP_015298898.1 27 - 28 3432 108
          XM_015443413.1 XP_015298899.1 27 - 28 3411 108
          XM_015443414.1 XP_015298900.1 28 - 29 3675 108

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Probe spans exons

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline