Search
Search

INTS8| Gene Symbol: INTS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563733.2 | XP_005563790.1 | 1 - 2 | 271 | 102 | |
| XM_005563734.2 | XP_005563791.1 | 1 - 2 | 276 | 102 | ||
Set Membership: |
|