Search
Search

MAST4| Gene Symbol: MAST4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557056.2 | XP_005557113.1 | 23 - 24 | 3166 | 109 | |
| XM_015451574.1 | XP_015307060.1 | 18 - 19 | 9878 | 109 | ||
| XM_015451576.1 | XP_015307062.1 | 19 - 20 | 2836 | 109 | ||
| XM_015451577.1 | XP_015307063.1 | 19 - 20 | 2832 | 109 | ||
| GenBank mRNA | AB056382.1 | - | 3 - 4 | 837 | 109 | |
Set Membership: |
|