Search
Search

ABCC5| Gene Symbol: ABCC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546512.2 | XP_005546569.1 | 14 - 15 | 2224 | 74 | |
| XM_005546513.2 | XP_005546570.1 | 14 - 15 | 2273 | 74 | ||
| XM_005546514.2 | XP_005546571.1 | 14 - 15 | 3442 | 74 | ||
| XM_005546515.2 | XP_005546572.1 | 10 - 11 | 5697 | 74 | ||
| XM_015445582.1 | XP_015301068.1 | 13 - 14 | 2321 | 74 | ||
Set Membership: |
|