Search
Search

PRG4| Gene Symbol: PRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540246.2 | XP_005540303.1 | 8 - 9 | 3327 | 74 | |
| XM_005540249.2 | XP_005540306.1 | 6 - 7 | 3106 | 74 | ||
| XM_015447161.1 | XP_015302647.1 | 8 - 9 | 3385 | 74 | ||
| XM_015447165.1 | XP_015302651.1 | 7 - 8 | 3204 | 74 | ||
| XM_015447171.1 | XP_015302657.1 | 7 - 8 | 3198 | 74 | ||
| XM_015447178.1 | XP_015302664.1 | 6 - 7 | 3048 | 74 | ||
| XM_015447182.1 | XP_015302668.1 | 5 - 6 | 2925 | 74 | ||
Set Membership: |
|