Search
Search

GRK5| Gene Symbol: GRK5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566565.2 | XP_005566622.1 | 10 - 11 | 1184 | 74 | |
| XM_005566566.2 | XP_005566623.1 | 10 - 11 | 1184 | 74 | ||
| XM_005566567.2 | XP_005566624.1 | 9 - 10 | 1106 | 74 | ||
| XM_015456768.1 | XP_015312254.1 | 9 - 10 | 941 | 74 | ||
| XM_015456769.1 | XP_015312255.1 | 9 - 10 | 1088 | 74 | ||
| XM_015456770.1 | XP_015312256.1 | 10 - 11 | 967 | 74 | ||
Set Membership: |
|