Search
Search

INPP5F| Gene Symbol: INPP5F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566579.2 | XP_005566636.1 | 16 - 17 | 2188 | 132 | |
| XM_005566581.1 | XP_005566638.1 | 2 - 3 | 364 | 132 | ||
| XM_015456771.1 | XP_015312257.1 | 16 - 17 | 1953 | 132 | ||
Set Membership: |
|