Search
Search

RAPH1| Gene Symbol: RAPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573992.2 | XP_005574049.1 | 13 - 14 | 1955 | 110 | |
| XM_005573993.2 | XP_005574050.1 | 13 - 14 | 3232 | 110 | ||
| XM_005573995.2 | XP_005574052.1 | 11 - 12 | 1795 | 110 | ||
| XM_005573997.2 | XP_005574054.1 | 13 - 14 | 1924 | 110 | ||
| XM_015432589.1 | XP_015288075.1 | 12 - 13 | 2419 | 110 | ||
| XM_015432590.1 | XP_015288076.1 | 11 - 12 | 2344 | 110 | ||
| XM_015432591.1 | XP_015288077.1 | 11 - 12 | 2338 | 110 | ||
| XM_015432592.1 | XP_015288078.1 | 10 - 11 | 2263 | 110 | ||
| XM_015432593.1 | XP_015288079.1 | 12 - 13 | 2419 | 110 | ||
| XM_015432594.1 | XP_015288080.1 | 12 - 13 | 2419 | 110 | ||
Set Membership: |
|