Search
Search

MMEL1| Gene Symbol: MMEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544996.2 | XP_005545053.1 | 6 - 7 | 701 | 64 | |
| XM_005544997.2 | XP_005545054.1 | 6 - 7 | 701 | 64 | ||
| XM_005545000.2 | XP_005545057.1 | 6 - 7 | 701 | 64 | ||
Set Membership: |
|