Search
Search

CDC42BPB| Gene Symbol: CDC42BPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562262.2 | XP_005562319.1 | 27 - 28 | 3625 | 76 | |
| XM_015454228.1 | XP_015309714.1 | 28 - 29 | 3676 | 76 | ||
| XM_015454229.1 | XP_015309715.1 | 28 - 29 | 3580 | 76 | ||
| XM_015454230.1 | XP_015309716.1 | 27 - 28 | 3598 | 76 | ||
| XM_015454231.1 | XP_015309717.1 | 26 - 27 | 3547 | 76 | ||
Set Membership: |
|