Search
Search

LRRC28| Gene Symbol: LRRC28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560591.2 | XP_005560648.1 | 3 - 4 | 454 | 69 | |
| XM_005560592.2 | XP_005560649.1 | 3 - 4 | 459 | 69 | ||
| XM_005560593.2 | XP_005560650.1 | 3 - 4 | 454 | 69 | ||
| XM_005560594.2 | XP_005560651.1 | 3 - 4 | 454 | 69 | ||
| XM_015453443.1 | XP_015308929.1 | 2 - 3 | 418 | 69 | ||
Set Membership: |
|