Search
Search

ZNF283| Gene Symbol: ZNF283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319641.1 | NP_001306570.1 | 1 - 2 | 691 | 123 | |
| XM_005589474.2 | XP_005589531.1 | 5 - 6 | 558 | 123 | ||
| XM_005589475.2 | XP_005589532.1 | 2 - 3 | 336 | 123 | ||
| XM_005589476.2 | XP_005589533.1 | 2 - 3 | 336 | 123 | ||
| XM_015441105.1 | XP_015296591.1 | 1 - 2 | 1013 | 123 | ||
| GenBank mRNA | AB066540.1 | - | 1 - 2 | 691 | 123 | |
Set Membership: |
|