Search
Search

TNPO1| Gene Symbol: TNPO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557124.2 | XP_005557181.1 | 20 - 21 | 2488 | 77 | |
| XM_005557125.2 | XP_005557182.1 | 20 - 21 | 2446 | 77 | ||
| XM_005557126.2 | XP_005557183.1 | 20 - 21 | 2449 | 77 | ||
| GenBank mRNA | AB170336.1 | - | 12 - 13 | 1282 | 77 | |
Set Membership: |
|