Search
Search

TCF7L2| Gene Symbol: TCF7L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005566438.2 | XP_005566495.1 | 12 - 13 | 4838 | 75 | |
| XM_015456677.1 | XP_015312163.1 | 11 - 12 | 1761 | 75 | ||
| XM_015456680.1 | XP_015312166.1 | 11 - 12 | 1758 | 75 | ||
| XM_015456681.1 | XP_015312167.1 | 11 - 12 | 1751 | 75 | ||
| XM_015456682.1 | XP_015312168.1 | 11 - 12 | 1744 | 75 | ||
| XM_015456683.1 | XP_015312169.1 | 11 - 12 | 1734 | 75 | ||
| XM_015456686.1 | XP_015312172.1 | 10 - 11 | 1710 | 75 | ||
| XM_015456687.1 | XP_015312173.1 | 10 - 11 | 1693 | 75 | ||
| XM_015456688.1 | XP_015312174.1 | 12 - 13 | 2926 | 75 | ||
| XM_015456689.1 | XP_015312175.1 | 13 - 14 | 2839 | 75 | ||
| XM_015456690.1 | XP_015312176.1 | 12 - 13 | 2923 | 75 | ||
| XM_015456691.1 | XP_015312177.1 | 10 - 11 | 1614 | 75 | ||
| XM_015456692.1 | XP_015312178.1 | 10 - 11 | 1599 | 75 | ||
| XM_015456694.1 | XP_015312180.1 | 10 - 11 | 1589 | 75 | ||
| XM_015456697.1 | XP_015312183.1 | 9 - 10 | 1548 | 75 | ||
| XM_015456698.1 | XP_015312184.1 | 11 - 12 | 2780 | 75 | ||
| XM_015456699.1 | XP_015312185.1 | 11 - 12 | 2770 | 75 | ||
| XM_015456700.1 | XP_015312186.1 | 11 - 12 | 2763 | 75 | ||
Set Membership: |
|