Search
Search

ITGAX| Gene Symbol: ITGAX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591741.2 | XP_005591798.2 | 21 - 22 | 2748 | 115 | |
| XM_015442665.1 | XP_015298151.1 | 21 - 22 | 2748 | 115 | ||
| XM_015442666.1 | XP_015298152.1 | 21 - 22 | 2748 | 115 | ||
| XM_015442667.1 | XP_015298153.1 | 20 - 21 | 2834 | 115 | ||
Set Membership: |
|