Search
Search

PFKFB2| Gene Symbol: PFKFB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283120.1 | NP_001270049.1 | 10 - 11 | 1133 | 90 | |
| XM_015442188.1 | XP_015297674.1 | 12 - 13 | 1354 | 90 | ||
| XM_015444841.1 | XP_015300327.1 | 12 - 13 | 1408 | 90 | ||
| XM_015445182.1 | XP_015300668.1 | 12 - 13 | 1418 | 90 | ||
| XM_015445472.1 | XP_015300958.1 | 13 - 14 | 1401 | 90 | ||
| XM_015445812.1 | XP_015301298.1 | 12 - 13 | 1312 | 90 | ||
| XM_015446225.1 | XP_015301711.1 | 12 - 13 | 1364 | 90 | ||
| XM_015446750.1 | XP_015302236.1 | 12 - 13 | 1354 | 90 | ||
| XM_015447144.1 | XP_015302630.1 | 12 - 13 | 1379 | 90 | ||
| XM_015447387.1 | XP_015302873.1 | 12 - 13 | 1354 | 90 | ||
| GenBank mRNA | AB179340.1 | - | 10 - 11 | 1134 | 90 | |
Set Membership: |
|