Search
Search

NCOA1| Gene Symbol: NCOA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576420.2 | XP_005576477.1 | 14 - 15 | 3134 | 86 | |
| XM_005576422.2 | XP_005576479.1 | 14 - 15 | 3134 | 86 | ||
| XM_005576423.2 | XP_005576480.1 | 14 - 15 | 3134 | 86 | ||
| XM_005576424.2 | XP_005576481.1 | 14 - 15 | 3134 | 86 | ||
| XM_015433940.1 | XP_015289426.1 | 13 - 14 | 3514 | 86 | ||
Set Membership: |
|