Search
Search

KTN1| Gene Symbol: KTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005561328.2 | XP_005561385.1 | 23 - 24 | 2642 | 110 | |
| XM_005561330.2 | XP_005561387.1 | 22 - 23 | 2493 | 110 | ||
| XM_005561331.2 | XP_005561388.1 | 23 - 24 | 2642 | 110 | ||
| XM_005561332.2 | XP_005561389.1 | 22 - 23 | 2501 | 110 | ||
| XM_005561334.2 | XP_005561391.1 | 22 - 23 | 2493 | 110 | ||
Set Membership: |
|