Search
Search

SLITRK2| Gene Symbol: SLITRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594753.2 | XP_005594810.1 | 5 | 3856 | 147 | |
| XM_005594754.2 | XP_005594811.1 | 5 | 3867 | 147 | ||
| XM_005594755.2 | XP_005594812.1 | 5 | 3845 | 147 | ||
| XM_015444229.1 | XP_015299715.1 | 5 | 4882 | 147 | ||
| XM_015444230.1 | XP_015299716.1 | 5 | 4893 | 147 | ||
| XM_015444231.1 | XP_015299717.1 | 5 | 4871 | 147 | ||
| GenBank mRNA | AB063047.1 | - | 1 | 1936 | 148 | |
Set Membership: |
|