Search
Search

RAB11FIP4| Gene Symbol: RAB11FIP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583365.2 | XP_005583422.1 | 5 - 6 | 872 | 73 | |
| XM_005583366.2 | XP_005583423.1 | 5 - 6 | 1446 | 73 | ||
| XM_015438147.1 | XP_015293633.1 | 7 - 8 | 886 | 73 | ||
Set Membership: |
|