Search
Search

OBSL1| Gene Symbol: OBSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574406.1 | XP_005574463.1 | 16 - 17 | 5159 | 146 | |
| XM_015432863.1 | XP_015288349.1 | 15 - 16 | 4883 | 146 | ||
| XM_015432865.1 | XP_015288351.1 | 15 - 16 | 4883 | 146 | ||
Set Membership: |
|