Search
Search

SLC27A4| Gene Symbol: SLC27A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284008.1 | NP_001270937.1 | 4 - 5 | 792 | 72 | |
| XM_015436097.1 | XP_015291583.1 | 6 - 7 | 1073 | 72 | ||
| XM_015436098.1 | XP_015291584.1 | 6 - 7 | 1078 | 72 | ||
| XM_015436099.1 | XP_015291585.1 | 5 - 6 | 1079 | 72 | ||
| GenBank mRNA | AB170334.1 | - | 3 - 4 | 632 | 72 | |
| AB179131.1 | - | 4 - 5 | 793 | 72 | ||
Set Membership: |
|