Search
Search

LRRC71| Gene Symbol: LRRC71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541386.2 | XP_005541443.1 | 5 - 6 | 792 | 56 | |
| XM_005541387.2 | XP_005541444.1 | 5 - 6 | 792 | 56 | ||
| XM_005541389.2 | XP_005541446.1 | 5 - 6 | 792 | 56 | ||
| XM_015455417.1 | XP_015310903.1 | 5 - 6 | 792 | 56 | ||
| XM_015455422.1 | XP_015310908.1 | 5 - 6 | 792 | 56 | ||
| XM_015455425.1 | XP_015310911.1 | 5 - 6 | 792 | 56 | ||
| XM_015455433.1 | XP_015310919.1 | 5 - 6 | 792 | 56 | ||
| XM_015455434.1 | XP_015310920.1 | 5 - 6 | 792 | 56 | ||
Set Membership: |
|