Search
Search

SLC6A9| Gene Symbol: SLC6A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543613.2 | XP_005543670.1 | 12 - 13 | 2957 | 68 | |
| XM_005543618.2 | XP_005543675.1 | 11 - 12 | 1799 | 68 | ||
| XM_005543620.2 | XP_005543677.1 | 12 - 13 | 1897 | 68 | ||
| XM_005543621.2 | XP_005543678.1 | 10 - 11 | 2637 | 68 | ||
| XM_005543626.2 | XP_005543683.1 | 10 - 11 | 2524 | 68 | ||
| XM_005543628.2 | XP_005543685.1 | 11 - 12 | 1741 | 68 | ||
| XM_005543629.2 | XP_005543686.1 | 11 - 12 | 1742 | 68 | ||
| XM_015437004.1 | XP_015292490.1 | 12 - 13 | 1899 | 68 | ||
Set Membership: |
|