Search
Search

EIF4ENIF1| Gene Symbol: EIF4ENIF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005567526.2 | XP_005567583.1 | 4 - 5 | 621 | 115 | |
| XM_005567527.2 | XP_005567584.1 | 4 - 5 | 506 | 115 | ||
| XM_005567528.2 | XP_005567585.1 | 4 - 5 | 506 | 115 | ||
Set Membership: |
|