Search
Search

IL34| Gene Symbol: IL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592494.2 | XP_005592551.1 | 6 - 7 | 1129 | 57 | |
| XM_005592495.2 | XP_005592552.1 | 6 - 7 | 1192 | 57 | ||
| XM_005592496.2 | XP_005592553.1 | 6 - 7 | 1126 | 57 | ||
| XM_015442832.1 | XP_015298318.1 | 6 - 7 | 1095 | 57 | ||
Set Membership: |
|