Search
Search

PLCE1| Gene Symbol: PLCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565986.2 | XP_005566043.1 | 24 - 25 | 5737 | 93 | |
| XM_005565988.2 | XP_005566045.1 | 23 - 24 | 5717 | 93 | ||
| XM_005565989.2 | XP_005566046.1 | 24 - 25 | 5689 | 93 | ||
| XM_005565990.2 | XP_005566047.1 | 23 - 24 | 5669 | 93 | ||
| XM_005565991.2 | XP_005566048.1 | 22 - 23 | 4321 | 93 | ||
| XM_015456402.1 | XP_015311888.1 | 23 - 24 | 5703 | 93 | ||
Set Membership: |
|