Search
Search

MGAT5| Gene Symbol: MGAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572984.2 | XP_005573041.1 | 16 - 17 | 2266 | 76 | |
| XM_005572985.2 | XP_005573042.1 | 16 - 17 | 2270 | 76 | ||
| XM_005572986.2 | XP_005573043.1 | 16 - 17 | 3951 | 76 | ||
| XM_005572987.2 | XP_005573044.1 | 16 - 17 | 3030 | 76 | ||
| XM_015432007.1 | XP_015287493.1 | 16 - 17 | 3954 | 76 | ||
| XM_015432008.1 | XP_015287494.1 | 16 - 17 | 3338 | 76 | ||
| XM_015432009.1 | XP_015287495.1 | 15 - 16 | 2164 | 76 | ||
Set Membership: |
|