Search
Search

PLA2R1| Gene Symbol: PLA2R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573281.2 | XP_005573338.2 | 27 - 28 | 4442 | 72 | |
| XM_015432273.1 | XP_015287759.1 | 27 - 28 | 4442 | 72 | ||
| XM_015432274.1 | XP_015287760.1 | 26 - 27 | 4268 | 72 | ||
| XM_015432275.1 | XP_015287761.1 | 25 - 26 | 3884 | 72 | ||
Set Membership: |
|