Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mf01075718_m1
          See other SEC31B GE Assays ›
          Gene Symbol:
          SEC31B
          Gene Name:
          SEC31 homolog B, COPII coat complex component
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 99417027 - 99452222 on Build M_fas_5.0
          Species:
          Macaca fascicularis
          Species Specific ID (Flybase ID):
          -
          Assay ID Mf01075718_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SEC31B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005566195.1 XP_005566252.1 19 - 20 2656 91
          XM_015456549.1 XP_015312035.1 22 - 23 3161 91
          XM_015456550.1 XP_015312036.1 22 - 23 3503 91
          XM_015456552.1 XP_015312038.1 22 - 23 4076 91
          XM_015456553.1 XP_015312039.1 22 - 23 3457 91
          XM_015456554.1 XP_015312040.1 22 - 23 3078 91
          XM_015456555.1 XP_015312041.1 22 - 23 3420 91
          XM_015456556.1 XP_015312042.1 22 - 23 3158 91
          XM_015456557.1 XP_015312043.1 22 - 23 3161 91
          XM_015456558.1 XP_015312044.1 21 - 22 3151 91
          XM_015456559.1 XP_015312045.1 22 - 23 3160 91
          XM_015456560.1 XP_015312046.1 21 - 22 3148 91
          XM_015456561.1 XP_015312047.1 21 - 22 4064 91
          XM_015456563.1 XP_015312049.1 21 - 22 3066 91
          XM_015456564.1 XP_015312050.1 22 - 23 3160 91
          XM_015456565.1 XP_015312051.1 22 - 23 3160 91
          XM_015456566.1 XP_015312052.1 21 - 22 3150 91
          XM_015456567.1 XP_015312053.1 21 - 22 3147 91
          XM_015456568.1 XP_015312054.1 22 - 23 3160 91
          XM_015456569.1 XP_015312055.1 21 - 22 2990 91
          XM_015456570.1 XP_015312056.1 21 - 22 2954 91
          XM_015456571.1 XP_015312057.1 21 - 22 3296 91
          XM_015456572.1 XP_015312058.1 21 - 22 2969 91
          XM_015456574.1 XP_015312060.1 18 - 19 2716 91
          XM_015456575.1 XP_015312061.1 18 - 19 2713 91
          XM_015456576.1 XP_015312062.1 19 - 20 3132 91
          XM_015456577.1 XP_015312063.1 21 - 22 2961 91
          XM_015456578.1 XP_015312064.1 20 - 21 2885 91

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline