Search
Search

UBAP2| Gene Symbol: UBAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581503.2 | XP_005581560.1 | 14 - 15 | 1668 | 56 | |
| XM_005581507.2 | XP_005581564.1 | 14 - 15 | 1668 | 56 | ||
| XM_015437076.1 | XP_015292562.1 | 13 - 14 | 1557 | 56 | ||
| XM_015437077.1 | XP_015292563.1 | 10 - 11 | 1263 | 56 | ||
Set Membership: |
|