Search
Search

RNF130| Gene Symbol: RNF130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558792.2 | XP_005558849.1 | 6 - 7 | 872 | 78 | |
| XM_005558794.2 | XP_005558851.2 | 6 - 7 | 875 | 78 | ||
| XM_005558795.2 | XP_005558852.1 | 5 - 6 | 800 | 78 | ||
| XM_005558796.2 | XP_005558853.1 | 6 - 7 | 875 | 78 | ||
| XM_015452375.1 | XP_015307861.1 | 6 - 7 | 875 | 78 | ||
Set Membership: |
|