Search
Search

FCER2| Gene Symbol: FCER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587775.2 | XP_005587832.1 | 9 - 10 | 864 | 60 | |
| XM_005587776.2 | XP_005587833.1 | 9 - 10 | 860 | 60 | ||
| XM_005587778.2 | XP_005587835.1 | 8 - 9 | 761 | 60 | ||
| XM_005587779.2 | XP_005587836.1 | 8 - 9 | 830 | 60 | ||
| XM_005587780.2 | XP_005587837.1 | 7 - 8 | 719 | 60 | ||
Set Membership: |
|