Search
Search

PHF8| Gene Symbol: PHF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593661.2 | XP_005593718.1 | 2 - 3 | 553 | 74 | |
| XM_005593663.2 | XP_005593720.1 | 2 - 3 | 906 | 74 | ||
| XM_005593664.2 | XP_005593721.1 | 2 - 3 | 857 | 74 | ||
| XM_005593665.2 | XP_005593722.1 | 2 - 3 | 1553 | 74 | ||
| XM_005593667.2 | XP_005593724.1 | 2 - 3 | 550 | 74 | ||
| XM_005593668.2 | XP_005593725.1 | 2 - 3 | 597 | 74 | ||
Set Membership: |
|