Search
Search

RBBP8| Gene Symbol: RBBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319568.1 | NP_001306497.1 | 2 - 3 | 154 | 154 | |
| XM_005587046.2 | XP_005587103.1 | 3 - 4 | 493 | 154 | ||
| XM_005587047.2 | XP_005587104.1 | 3 - 4 | 493 | 154 | ||
| XM_015440112.1 | XP_015295598.1 | 3 - 4 | 433 | 154 | ||
| XM_015440113.1 | XP_015295599.1 | 4 - 5 | 601 | 154 | ||
| GenBank mRNA | HM486765.1 | - | 2 - 3 | 154 | 154 | |
Set Membership: |
|